Pyridazine-3-carbonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Cyanopyridazine
OR48248-1G
OR48248-100MG
APOSOR48248-1GEA
58
EUR
APOSOR48248-1G
APOSOR48248-100MG
Pyridazine-3-carbonitrile 95%
Pyridazine-3-carbonitrile
Formel:
C₅H₃N₃ Molecular Weight: 105,1 g/mol Schmelzpunkt: 43…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09881239 CAS-Nummer: 53896-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...