2-Amino-6-chloropyridine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-6-chloroisonicotinic acid

OR48249-1G OR48249-250MG OR48249-5G
APOSOR48249-1GEA 284 EUR
APOSOR48249-1G APOSOR48249-250MG APOSOR48249-5G
2-Amino-6-chloropyridine-4-carboxylic acid
2-Amino-6-chloropyridine-4-carboxylic acid
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11036352
CAS-Nummer: 6313-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR