5-Nitroindole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Nitro-1H-indole-2-carboxylic acid

OR48250-1G OR48250-250MG
APOSOR48250-1GEA 24.8 EUR
APOSOR48250-1G APOSOR48250-250MG
5-Nitroindole-2-carboxylic acid
5-Nitroindole-2-carboxylic acid
Formel: C₉H₆N₂O₄
Molecular Weight: 206,157 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236372
CAS-Nummer: 16730-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR