Methyl-4-ethinylbenzoat 96%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-ethynylbenzoate
OR48275-1G
OR48275-25G
OR48275-5G
APOSOR48275-1GEA
31.9
EUR
APOSOR48275-1G
APOSOR48275-25G
APOSOR48275-5G
Methyl-4-ethinylbenzoat 96%
Methyl-4-ethinylbenzoat
Formel:
C₁₀H₈O₂ Molecular Weight: 160.17 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00168820 CAS-Nummer: 3034-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...