3,4-Dihydroquinolin-2(1H)-one 97%

Lieferant: Apollo Scientific
Achtung

OR4829-25G OR4829-5G
APOSOR4829-25GEA 72.5 EUR
APOSOR4829-25G APOSOR4829-5G
3,4-Dihydroquinolin-2(1H)-one 97%
3,4-Dihydroquinolin-2(1H)-one
Formel: C9H9NO
Molecular Weight: 147,18 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016722
CAS-Nummer: 553-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR