5-(2-Chlorphenyl)oxazol 95%

Lieferant: Apollo Scientific
Achtung

OR4832-1G OR4832-250MG
APOSOR4832-1GEA 139 EUR
APOSOR4832-1G APOSOR4832-250MG
5-(2-Chlorphenyl)oxazol 95%
5-(2-Chlorophenyl)oxazole
Formel: C₉H₆ClNO
Molecular Weight: 179,6030 g/mol
Schmelzpunkt: 33…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084973
CAS-Nummer: 89808-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR