2',4',6'-Trihydroxyacetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Phloroacetophenone, THAP, 2-Acetylphloroglucinol

OR4838-25G OR4838-5G
APOSOR4838-25GEA 43.5 EUR
APOSOR4838-25G APOSOR4838-5G
2',4',6'-Trihydroxyacetophenon 98%
2',4',6'-Trihydroxyacetophenon
Formel: C₈H₈O₄
Molecular Weight: 168,15 g/mol
Schmelzpunkt: 219…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149091
CAS-Nummer: 480-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR