2-Thenoesäurehydrazid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Thiophencarbonsäurehydrazid, Thiophen-2-carbonsäurehydrazid, Thiophen-2-carbohydrazid

OR4847-1G OR4847-25G
APOSOR4847-1GEA 21.9 EUR
APOSOR4847-1G APOSOR4847-25G
2-Thenoesäurehydrazid 98%
2-Thenoesäurehydrazid
Formel: C₅H₆N₂OS
Molecular Weight: 142,18 g/mol
Schmelzpunkt: 136…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005435
CAS-Nummer: 2361-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR