N-Benzoylthiourea 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Carbamothioylbenzamide, N-(Carbamothioyl)benzamide
OR4861-25G
OR4861-100G
OR4861-1G
APOSOR4861-25GEA
120
EUR
APOSOR4861-25G
APOSOR4861-100G
APOSOR4861-1G
N-Benzoylthiourea 98%
N-Benzoylthiourea
Formel:
C₈H₈N₂OS Molecular Weight: 180,23 g/mol Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041191 CAS-Nummer: 614-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...