3,5-Dichlorthiobenzamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dichlorthiobenzoesäureamid
OR49005-1G
OR49005-5G
APOSOR49005-1GEA
161
EUR
APOSOR49005-1G
APOSOR49005-5G
3,5-Dichlorthiobenzamid
3,5-Dichlorthiobenzamid
Formel:
C₇H₅Cl₂NS Molecular Weight: 206.09 g/mol Schmelzpunkt: 102…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973322 CAS-Nummer: 22179-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...