2-Chloro-1-(1H-indol-3-yl)ethanone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3CAI, 3-(Chloroacetyl)-1H-indole
OR49019-5G
OR49019-1G
OR49019-250MG
APOSOR49019-5GEA
1040
EUR
APOSOR49019-5G
APOSOR49019-1G
APOSOR49019-250MG
2-Chloro-1-(1H-indol-3-yl)ethanone 95%
2-Chloro-1-(1H-indol-3-yl)ethanone
Formel:
C₁₀H₈ClNO Lagertemperatur: Gefrierschrank |
MDL:
MFCD00464796 CAS-Nummer: 28755-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...