1-(3-Cyanphenyl)piperazin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Piperazin-1-yl)benzonitrile
OR49032-1G
OR49032-5G
APOSOR49032-1GEA
68.3
EUR
APOSOR49032-1G
APOSOR49032-5G
1-(3-Cyanphenyl)piperazin 95%
1-(3-Cyanphenyl)piperazin
Formel:
C₁₁H₁₃N₃ Molecular Weight: 187,2410 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06796181 CAS-Nummer: 178928-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...