4-(Phenylethynyl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR49037-5G
APOSOR49037-5GEA 231 EUR
APOSOR49037-5G
4-(Phenylethynyl)benzoic acid 97%
4-(Phenylethynyl)benzoic acid
Formel: C₁₅H₁₀O₂
Molecular Weight: 222,25 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00438677
CAS-Nummer: 25739-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR