4,4'-Diaminodiphenylamine sulphate Hydrate 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: N1-(4-Aminophenyl)benzene-1,4-diamine sulphate, Bis(4-aminophenyl)amine Sulphate, 4,4'-Iminodianiline sulphate

OR4904-25G OR4904-5G
APOSOR4904-25GEA 29 EUR
APOSOR4904-25G APOSOR4904-5G
4,4'-Diaminodiphenylamine sulphate Hydrate 97+%
4,4'-Diaminodiphenylamine sulphate Hydrate
Formel: C₁₂H₁₃N₃
Molecular Weight: 297,33 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013113
CAS-Nummer: 53760-27-3
EINECS: 258-748-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR