2-Thioxo-4-imidazolidinon
Lieferant: Apollo Scientific
Synonyme:
2-Thioxoimidazolidin-4-one
OR49042-25G
APOSOR49042-25GEA
133
EUR
APOSOR49042-25G
2-Thioxo-4-imidazolidinon
2-Thiohydantoin
Formel:
C₃H₄N₂OS Molecular Weight: 116,14 g/mol Schmelzpunkt: 229…231 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005277 CAS-Nummer: 503-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...