H-D-4-Pal-OH·2HCl 97%
Lieferant: Apollo Scientific
Synonyme: (R)-2-Amino-3-(4-pyridyl)propionic acid hydrochlorate, 3-(4-Pyridyl)-D-alanine hydrochlorate, 3-(4-Pyridyl)-D-alanine·2HCl, H-D-4-Pal-OH.2HCl, D-4-Pyridylalanine hydrochlorate
OR49075-1G OR49075-250MG OR49075-5G
APOSOR49075-1GEA 69.8 EUR
APOSOR49075-1G APOSOR49075-250MG APOSOR49075-5G
H-D-4-Pal-OH·2HCl 97%
H-D-4-Pal-OH·2HCl
Formel: C₈H₁₀N₂O₂·₂HCl
Molecular Weight: 239,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09842075
CAS-Nummer: 174096-41-4