3,5-Dimethoxybenzoesäure 97%

Lieferant: Apollo Scientific
Achtung

OR4991-500G
APOSOR4991-500GEA 224 EUR
APOSOR4991-500G
3,5-Dimethoxybenzoesäure 97%
3,5-Dimethoxybenzoic acid
Formel: C₉H₁₀O₄
Molecular Weight: 182,18 g/mol
Schmelzpunkt: 181…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002502
CAS-Nummer: 1132-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR