3-Iodbenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
m-Iodobenzoic acid
OR4998-100G
OR4998-25G
APOSOR4998-100GEA
79.9
EUR
APOSOR4998-100G
APOSOR4998-25G
3-Iodbenzoesäure 98%
3-Iodbenzoesäure
Formel:
C₇H₅IO₂ Molecular Weight: 248,02 g/mol Schmelzpunkt: 185…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002496 CAS-Nummer: 618-51-9 EINECS: 210-555-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...