4-Hydroxybenzophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
p-Hydroxybenzophenon
OR5011-100G
OR5011-500G
OR5011-1KG
APOSOR5011-100GEA
31.9
EUR
APOSOR5011-100G
APOSOR5011-500G
APOSOR5011-1KG
4-Hydroxybenzophenon 98%
4-Hydroxybenzophenon
Formel:
C₁₃H₁₀O₂ Molecular Weight: 198.22 g/mol Siedepunkt: 260…262 °C/24mmHg Schmelzpunkt: 132…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002355 CAS-Nummer: 1137-42-4 EINECS: 214-507-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...