5-Bromo-[2,2']bipyridinyl

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2,2'-bipyridine

OR50952-1G OR50952-250MG OR50952-5G
APOSOR50952-1GEA 78.3 EUR
APOSOR50952-1G APOSOR50952-250MG APOSOR50952-5G
5-Bromo-[2,2']bipyridinyl
5-Bromo[2,2']bipyridinyl
Formel: C₁₀H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08234848
CAS-Nummer: 15862-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR