1,3-Dimethyl-1H-pyrazole-4-carbohydrazide 95%
Lieferant: Apollo Scientific
OR50953-1G
OR50953-250MG
APOSOR50953-1GEA
625
EUR
APOSOR50953-1G
APOSOR50953-250MG
1,3-Dimethyl-1H-pyrazole-4-carbohydrazide 95%
1,3-Dimethyl-1H-pyrazole-4-carbohydrazide
Formel:
C₆H₁₀N₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419412 CAS-Nummer: 512810-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...