3-Bromodibenzofuran
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromodibenzo[b,d]furan
OR50961-1G
OR50961-250MG
OR50961-5G
APOSOR50961-1GEA
29
EUR
APOSOR50961-1G
APOSOR50961-250MG
APOSOR50961-5G
3-Bromodibenzofuran
3-Bromodibenzofuran
Formel:
C₁₂H₇BrO Molecular Weight: 247,09 g/mol Schmelzpunkt: 122…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09966042 CAS-Nummer: 26608-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...