2,2'-Dibromo-9,9'-spirobi[9H-fluorene] 99%
Lieferant: Apollo Scientific
Synonyme:
2,2'-Dibromo-9,9'-spirobi[fluorene]
OR51004-1G
OR51004-5G
OR51004-25G
APOSOR51004-1GEA
21.9
EUR
APOSOR51004-1G
APOSOR51004-5G
APOSOR51004-25G
2,2'-Dibromo-9,9'-spirobi[9H-fluorene] 99%
2,2'-Dibromo-9,9'-spirobi[9H-fluorene]
Formel:
C₂₅H₁₄Br₂ Molecular Weight: 474,20 g/mol Schmelzpunkt: 240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08704219 CAS-Nummer: 67665-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...