4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane 99%

Lieferant: Apollo Scientific

Synonyme: Triphenylen-2-ylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)triphenylene

OR51015-5G
APOSOR51015-5GEA 79.9 EUR
APOSOR51015-5G
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane 99%
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane
Formel: C₂₄H₂₃BO₂
Molecular Weight: 354,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26401844
CAS-Nummer: 890042-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR