4-(1-Pyrenyl)phenylboronic acid 95%

Lieferant: Apollo Scientific

Synonyme: 4-(1-Pyrenyl)benzeneboronic acid

OR51021-1G OR51021-5G
APOSOR51021-1GEA 109 EUR
APOSOR51021-1G APOSOR51021-5G
4-(1-Pyrenyl)phenylboronic acid 95%
4-(1-Pyrenyl)phenylboronic acid
Formel: C₂₂H₁₅BO₂
Molecular Weight: 322.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28130386
CAS-Nummer: 872050-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR