(4-(Hydroxymethyl)phenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-(Hydroxymethyl)phenyl)acetic acid, 4-(Hydroxymethyl)phenylacetic acid
OR5112-5G
OR5112-25G
OR5112-100G
APOSOR5112-5GEA
36.4
EUR
APOSOR5112-5G
APOSOR5112-25G
APOSOR5112-100G
(4-(Hydroxymethyl)phenyl)essigsäure 98%
(4-(Hydroxymethyl)phenyl)essigsäure
Formel:
C₉H₁₀O₃ Schmelzpunkt: 131…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065692 CAS-Nummer: 73401-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...