3-Chloro-6-methoxy-1-benzothiophene-2-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-6-methoxybenzo[b]thiophene-2-carboxylic acid

OR51179-1G OR51179-250MG
APOSOR51179-1GEA 224 EUR
APOSOR51179-1G APOSOR51179-250MG
3-Chloro-6-methoxy-1-benzothiophene-2-carboxylic acid 95%
3-Chloro-6-methoxy-1-benzothiophene-2-carboxylic acid
Formel: C₁₀H₇ClO₃S
Molecular Weight: 242,68 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00448910
CAS-Nummer: 310390-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR