5-Chloroxindol 95%
Lieferant: Apollo Scientific
Synonyme:
5-Chloro-2-oxindole
OR51359-1G
OR51359-25G
OR51359-5G
APOSOR51359-1GEA
21.9
EUR
APOSOR51359-1G
APOSOR51359-25G
APOSOR51359-5G
5-Chloroxindol 95%
5-Chloroxindol
Formel:
C₈H₆ClNO Molecular Weight: 167,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191927 CAS-Nummer: 17630-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...