1-tert-Butylpiperazine 98+%
Lieferant: Apollo Scientific
|
Achtung
|
OR5147-1G
OR5147-5G
APOSOR5147-1GEA
31.9
EUR
APOSOR5147-1G
APOSOR5147-5G
1-tert-Butylpiperazine 98+%
1-tert-Butylpiperazine
Formel:
C₈H₁₈N₂ Molecular Weight: 142,24 g/mol Siedepunkt: 66…75 °C/12mmHg Schmelzpunkt: 32…40 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00049359 CAS-Nummer: 38216-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...