3-Chlor-2-formylpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloropyridine-2-carboxaldehyde

OR51758-1G OR51758-250MG
APOSOR51758-1GEA 66.7 EUR
APOSOR51758-1G APOSOR51758-250MG
3-Chlor-2-formylpyridin
3-Chlor-2-formylpyridin
Formel: C₆H₄ClNO
Molecular Weight: 141,56 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09991855
CAS-Nummer: 206181-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR