5-Brom-2-iodanilin

Lieferant: Apollo Scientific
Achtung

OR51804-1G OR51804-25G OR51804-5G
APOSOR51804-1GEA 29 EUR
APOSOR51804-1G APOSOR51804-25G APOSOR51804-5G
5-Brom-2-iodanilin
5-Brom-2-iodanilin
Formel: C₆H₅BrIN
Molecular Weight: 297,9200 g/mol
Schmelzpunkt: 55 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09753734
CAS-Nummer: 64085-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR