4-(Diphenylphosphino)benzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR51829-5G
APOSOR51829-5GEA
136
EUR
APOSOR51829-5G
4-(Diphenylphosphino)benzoesäure
4-(Diphenylphosphino)benzoic acid
Formel:
C₁₉H₁₅O₂P Molecular Weight: 306,3 g/mol Schmelzpunkt: 157…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00407264 CAS-Nummer: 2129-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...