2-Brom-3-chloranilin 98%

Lieferant: Apollo Scientific
Gefahr

OR51860-5G OR51860-1G OR51860-25G
APOSOR51860-5GEA 31.9 EUR
APOSOR51860-5G APOSOR51860-1G APOSOR51860-25G
2-Brom-3-chloranilin 98%
2-Brom-3-chloranilin
Formel: C₆H₅BrClN
Molecular Weight: 206,4700 g/mol
Schmelzpunkt: 41…43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10699543
CAS-Nummer: 96558-73-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR