Methyl-4-brom-2-methylbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-bromo-2-methylbenzoate

OR51873-1G OR51873-25G OR51873-5G
APOSOR51873-1GEA 49.3 EUR
APOSOR51873-1G APOSOR51873-25G APOSOR51873-5G
Methyl-4-brom-2-methylbenzoat
Methyl-4-brom-2-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,0706 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09954957
CAS-Nummer: 99548-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR