8-Hydroxy-7-chinolincarbonsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Hydroxy-7-quinolinecarboxylic acid
OR51895-1G
OR51895-5G
APOSOR51895-1GEA
34.8
EUR
APOSOR51895-1G
APOSOR51895-5G
8-Hydroxy-7-chinolincarbonsäure 95%
8-Hydroxy-7-chinolincarbonsäure
Formel:
C₁₀H₇NO₃ Molecular Weight: 189,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671539 CAS-Nummer: 19829-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...