2-(1H-Imidazol-4-ylessigsäurehydrochlorid) 95%

Lieferant: Apollo Scientific

Synonyme: 4-Imidazoleacetic acid hydrochloride, Imidazole-4-acetic acid monohydrochloride

OR51912-1G OR51912-250MG
APOSOR51912-1GEA 23.2 EUR
APOSOR51912-1G APOSOR51912-250MG
2-(1H-Imidazol-4-ylessigsäurehydrochlorid) 95%
2-(1H-Imidazol-5-yl)acetic acid hydrochloride
Formel: C₅H₇ClN₂O₂
Schmelzpunkt: 218…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012698
CAS-Nummer: 3251-69-2
EINECS: 221-840-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR