4-Nitro-m-anissäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxy-4-nitrobenzoic acid
OR5192-100G
OR5192-25G
OR5192-5G
APOSOR5192-100GEA
109
EUR
APOSOR5192-100G
APOSOR5192-25G
APOSOR5192-5G
4-Nitro-m-anissäure
4-Nitro-m-anissäure
Formel:
C₈H₇NO₅ Molecular Weight: 197,15 g/mol Schmelzpunkt: 233…235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007353 CAS-Nummer: 5081-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...