N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl N,N'-Bis(tert-butoxycarbonyl)carbamimidothioate, N,N'-Di-Boc-carbamimidothioic Acid Methyl Ester, 1,3-Di-Boc-2-methylisothiourea, N,N'-Bis(tert-butoxycarbonyl)carbamimidothioic Acid Methyl Ester, N,N'-Di-Boc-S-methylisothiourea, Methyl N,N'-Di-Boc-carbamimidothioate, 1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea
OR51930-100G
OR51930-25G
OR51930-5G
APOSOR51930-100GEA
199
EUR
APOSOR51930-100G
APOSOR51930-25G
APOSOR51930-5G
N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea 98%
N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea
Formel:
C₁₂H₂₂N₂O₄S Schmelzpunkt: 115…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00239356 CAS-Nummer: 107819-90-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...