3-Brombenzylmercaptan

Lieferant: Apollo Scientific
Achtung

Synonyme: m-Bromotoluene-α-thiol

OR51962-1G OR51962-5G
APOSOR51962-1GEA 26.1 EUR
APOSOR51962-1G APOSOR51962-5G
3-Brombenzylmercaptan
3-Brombenzylmercaptan
Formel: C₇H₇BrS
Molecular Weight: 203,1 g/mol
Flammpunkt: 120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06798010
CAS-Nummer: 886497-84-3
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR