Ethyl-5-bromo-2-methylnicotinate 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-bromo-2-methylnicotinate

OR51975-100G OR51975-25G OR51975-5G
APOSOR51975-100GEA 1860 EUR
APOSOR51975-100G APOSOR51975-25G APOSOR51975-5G
Ethyl-5-bromo-2-methylnicotinate 97+%
Ethyl-5-bromo-2-methylnicotinate
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698571
CAS-Nummer: 129477-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR