tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate 95%
Lieferant: Apollo Scientific
Synonyme: 2,6-Diaza-spiro[3.3]heptane-2-carboxylic acid tert-butyl ester oxalate salt, tert-Butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate
OR51986-1G OR51986-250MG OR51986-5G
APOSOR51986-1GEA 139 EUR
APOSOR51986-1G APOSOR51986-250MG APOSOR51986-5G
tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate 95%
tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate
Formel: C₁₂H₂₀N₂O₆
Molecular Weight: 288.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11976177
CAS-Nummer: 1227382-01-5