Methyl-4'-brombiphenyl-4-carboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4'-bromo[1,1'-biphenyl]-4-carboxylate, Methyl 4'-bromo-[1,1'-biphenyl]-4-carboxylate
OR52003-10G
OR52003-1G
OR52003-5G
APOSOR52003-10GEA
778
EUR
APOSOR52003-10G
APOSOR52003-1G
APOSOR52003-5G
Methyl-4'-brombiphenyl-4-carboxylat 95%
Methyl-4'-brombiphenyl-4-carboxylat
Formel:
C₁₄H₁₁BrO₂ Molecular Weight: 291.14 g/mol Schmelzpunkt: 140…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00496638 CAS-Nummer: 89901-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...