4-Formyl-2-methylthiazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-1,3-thiazole-4-carboxaldehyde, 2-Methylthiazole-4-carbaldehyde
OR5201-1G
OR5201-250MG
APOSOR5201-1GEA
29
EUR
APOSOR5201-1G
APOSOR5201-250MG
4-Formyl-2-methylthiazole 97%
4-Formyl-2-methylthiazole
Formel:
C₅H₅NOS Molecular Weight: 127,1643 g/mol Schmelzpunkt: 56…58 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06655197 CAS-Nummer: 20949-84-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...