4-Brom-4'-methoxybiphenyl 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-4'-methoxy-1,1'-biphenyl, 1-Bromo-4-(4-methoxyphenyl)benzene

OR52012-5G
APOSOR52012-5GEA 55.1 EUR
APOSOR52012-5G
4-Brom-4'-methoxybiphenyl 98%
4-Brom-4'-methoxybiphenyl
Formel: C₁₃H₁₁BrO
Molecular Weight: 263,1298 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00671954
CAS-Nummer: 58743-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR