Methyl-2,4-dimethoxypyrimidine-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2,4-dimethoxypyrimidine-5-carboxylate

OR52034-1G OR52034-250MG OR52034-5G
APOSOR52034-1GEA 111 EUR
APOSOR52034-1G APOSOR52034-250MG APOSOR52034-5G
Methyl-2,4-dimethoxypyrimidine-5-carboxylate
Methyl-2,4-dimethoxypyrimidine-5-carboxylate
Formel: C₈H₁₀N₂O₄
Molecular Weight: 198.1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11617311
CAS-Nummer: 15400-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR