6-(1-Pyrrolidinyl)-3-pyridinamine
Lieferant: Apollo Scientific
Synonyme:
5-Amino-2-(pyrrolidin-1-yl)pyridine
OR52050-1G
OR52050-100MG
OR52050-250MG
OR52050-500MG
OR52050-5G
APOSOR52050-1GEA
275
EUR
APOSOR52050-1G
APOSOR52050-100MG
APOSOR52050-250MG
APOSOR52050-500MG
APOSOR52050-5G
6-(1-Pyrrolidinyl)-3-pyridinamine
6-(1-Pyrrolidinyl)-3-pyridinamine
Formel:
C₉H₁₃N₃ Molecular Weight: 163,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425745 CAS-Nummer: 92808-19-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...