8-Aminoimidazo[1,2-a]pyridin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Imidazo[1,2-a]pyridin-8-ylamine
OR52051-1G
OR52051-100MG
OR52051-5G
APOSOR52051-1GEA
125
EUR
APOSOR52051-1G
APOSOR52051-100MG
APOSOR52051-5G
8-Aminoimidazo[1,2-a]pyridin 95%
8-Aminoimidazo[1,2-a]pyridin
Formel:
C₇H₇N₃ Molecular Weight: 133.1500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739233 CAS-Nummer: 73221-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...