Diethyl iminodiacetate 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Iminodiacetic Acid Diethyl Ester, Diethyl 2,2'-azanediyldiacetate
OR52082-250G
OR52082-50G
OR52082-25G
OR52082-100G
APOSOR52082-250GEA
69.8
EUR
APOSOR52082-250G
APOSOR52082-50G
APOSOR52082-25G
APOSOR52082-100G
Diethyl iminodiacetate 90%
Diethyl iminodiacetate
Formel:
C₈H₁₅NO₄ Molecular Weight: 189,21 g/mol Siedepunkt: 208 °C Dichte: 1,056 g/cm³ (25 °C) Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041925 CAS-Nummer: 6290-05-7 EINECS: 228-533-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...