Methyl-1-benzyl-3-pyrrolidincarboxylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1-benzylpyrrolidine-3-carboxylate

OR52084-1G OR52084-5G
APOSOR52084-1GEA 65.5 EUR
APOSOR52084-1G APOSOR52084-5G
Methyl-1-benzyl-3-pyrrolidincarboxylat 98%
Methyl-1-benzylpyrrolidine-3-carboxylate
Formel: C₁₃H₁₇NO₂
Molecular Weight: 219.28 g/mol
Siedepunkt: 98 °C/0.2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04038678
CAS-Nummer: 17012-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR