Methyl-1-benzylpiperidine-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1-benzylpiperidine-4-carboxylate

OR52086-5G OR52086-10G OR52086-25G
APOSOR52086-5GEA 26.1 EUR
APOSOR52086-5G APOSOR52086-10G APOSOR52086-25G
Methyl-1-benzylpiperidine-4-carboxylate
Methyl-1-benzylpiperidine-4-carboxylate
Formel: C₁₄H₁₉NO₂
Molecular Weight: 233.3100 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09750949
CAS-Nummer: 10315-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR